CH6371380609 Chem Impex International, Inc.

L-Leucyl-L-phenylalanine methylamide

  • MDL number MFCD01862383
  • Cas Number : 169812-43-5
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-leucyl-l-phenylalanine-methylamide/5680?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-leucyl-l-phenylalanine-methylamide/5680?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart