CH6371380618 Chem Impex International, Inc.

L-1,2,3,4-Tetrahydronorharman-3-carboxylic acid methyl ester hydrochloride

  • MDL number MFCD10042201
  • Cas Number : 83159-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242-246
Synonym L-Tpi-OMe HCl
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white flakes
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-1-2-3-4-tetrahydronorharman-3-carboxylic-acid-methyl-ester-hydrochloride/5692?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-1-2-3-4-tetrahydronorharman-3-carboxylic-acid-methyl-ester-hydrochloride/5692?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart