CH6371380620 Chem Impex International, Inc.

L-3,5-Diiodotyrosine ethyl ester hydrochloride

  • MDL number MFCD00035073

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-207
Synonym 3,5-Diiodo-L-Tyr-OEt�
Purity 97
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-3-5-diiodotyrosine-ethyl-ester-hydrochloride/5694?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-3-5-diiodotyrosine-ethyl-ester-hydrochloride/5694?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart