CH6371380621 Chem Impex International, Inc.

O-Allyl-L-tyrosine methyl ester hydrochloride

  • MDL number MFCD01862374
  • Cas Number : 138535-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-194
Synonym L-Tyr(All)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-allyl-l-tyrosine-methyl-ester-hydrochloride/5695?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-allyl-l-tyrosine-methyl-ester-hydrochloride/5695?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart