CH6371380628 Chem Impex International, Inc.

(R)-(-)-2-(Methoxymethyl)pyrrolidine

  • MDL number MFCD00066219
  • Cas Number : 84025-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99.5
Density 0.932 g/mL at 20
Storage temp. Store at 0-8 °C
Physical Form Colorless liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-methyl-d-prolinol/5707?cid=0

Safety Information

hcodes 3
MSDS http://www.chemimpex.com/product/productinfo/i-o-i-methyl-d-prolinol/5707?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart