CH6371380630 Chem Impex International, Inc.

Phthaloyl-DL-glutamic acid

  • MDL number MFCD00023059

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Pht-DL-Glu-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phthaloyl-dl-glutamic-acid/5712?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phthaloyl-dl-glutamic-acid/5712?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart