CH6371380635 Chem Impex International, Inc.

Phthaloyl-glycine 4-nitorophenyl ester

  • MDL number MFCD00190995
  • Cas Number : 21313-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-179 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phthaloyl-glycine-4-nitorophenyl-ester/5718?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phthaloyl-glycine-4-nitorophenyl-ester/5718?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart