CH6371380638 Chem Impex International, Inc.

Z-b-alanine amide

  • MDL number MFCD00270463
  • Cas Number : 886-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-165 �C
Synonym Z-beta-Ala-NH2
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-beta-alanine-amide/5723?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-beta-alanine-amide/5723?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart