CH6371380640 Chem Impex International, Inc.

Z-DL-4-amino-3-hydroxybutyric acid

  • MDL number MFCD00191057
  • Cas Number : 118125-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-dl-4-amino-3-hydroxybutyric-acid/5725?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-dl-4-amino-3-hydroxybutyric-acid/5725?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart