CH6371380642 Chem Impex International, Inc.

Na-Z-L-arginine benzyl ester

  • MDL number MFCD01862300
  • Cas Number : 103305-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Arg-OBzl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-arginine-benzyl-ester/5727?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-arginine-benzyl-ester/5727?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart