CH6371380645 Chem Impex International, Inc.

Z-L-aspartic acid-di-tert-butyl ester

  • MDL number MFCD00270579
  • Cas Number : 42417-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Asp(OtBu)-OtBu
Purity 98
Storage temp. Store at 0-8
Physical Form Colorless oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-di-i-tert-i-butyl-ester/5730?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-di-i-tert-i-butyl-ester/5730?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart