CH6371380646 Chem Impex International, Inc.

Z-L-2-cyclohexylglycine

  • MDL number MFCD00191083
  • Cas Number : 69901-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Chg-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-2-cyclohexylglycine/5731?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-2-cyclohexylglycine/5731?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart