CH6371380648 Chem Impex International, Inc.

Na-Z-Nd-xanthyl-D-glutamine

  • MDL number MFCD01862303
  • Cas Number : 327981-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-184
Synonym Z-D-Gln(Xan)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-delta-sup-xanthyl-d-glutamine/5733?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-delta-sup-xanthyl-d-glutamine/5733?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart