CH6371380649 Chem Impex International, Inc.

Na-Z-DL-histidine

  • MDL number MFCD00037852
  • Cas Number : 19728-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-DL-His-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-dl-histidine/5735?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-dl-histidine/5735?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart