CH6371380652 Chem Impex International, Inc.

Z-DL-norleucine

  • MDL number MFCD00037271
  • Cas Number : 15027-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-107
Synonym Z-DL-Nle-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-dl-norleucine/5738?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-dl-norleucine/5738?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart