CH6371380654 Chem Impex International, Inc.

Z-4-chloro-DL-phenylalanine

  • MDL number MFCD00191079
  • Cas Number : 55478-54-1
  • Molecular Weight : 333.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-163 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-4-chloro-dl-phenylalanine/5740?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-4-chloro-dl-phenylalanine/5740?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart