CH6371380656 Chem Impex International, Inc.

Na-Z-Nd-xanthyl-L-glutamine

  • MDL number MFCD01862302
  • Cas Number : 327981-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-185
Synonym Z-L-Gln(Xan)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-delta-sup-xanthyl-l-glutamine/5742?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-delta-sup-xanthyl-l-glutamine/5742?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart