CH6371380658 Chem Impex International, Inc.

Z-L-glutamic acid g-tert-butyl ester a-methyl ester

  • MDL number MFCD01862315
  • Cas Number : 56877-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Glu(OtBu)-OMe
Purity 98
Storage temp. Store at 0-8
Physical Form Viscous liquid to white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-methyl-ester/5744?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-methyl-ester/5744?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart