CH6371380660 Chem Impex International, Inc.

Z-L-leucine amide

  • MDL number MFCD00080252
  • Cas Number : 4801-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-123
Synonym Z-L-Leu-NH2
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-leucine-amide/5746?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-leucine-amide/5746?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart