CH6371380663 Chem Impex International, Inc.

Z-L-methionine N-hydroxysuccinimide ester

  • MDL number MFCD00190662
  • Cas Number : 3392-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-105
Synonym Z-L-Met-OSu
Purity 96
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-methionine-i-n-i-hydroxysuccinimide-ester/5749?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-methionine-i-n-i-hydroxysuccinimide-ester/5749?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart