CH6371380666 Chem Impex International, Inc.

Z-L-phenylalanine 4-nitrobenzyl ester

  • MDL number MFCD01076131

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-111
Synonym Z-L-Phe-ONb
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-4-nitrobenzyl-ester/5752?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-4-nitrobenzyl-ester/5752?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart