CH6371380667 Chem Impex International, Inc.

Z-L-Phenylalanine tert-butyl ester

  • MDL number MFCD00191137
  • Cas Number : 7670-20-4
  • Molecular Weight : 355.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-81
Purity 98
Storage temp. Store at 0-8
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-i-tert-i-butyl-ester/5753?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-i-tert-i-butyl-ester/5753?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart