CH6371380668 Chem Impex International, Inc.

Z-L-serine amide

  • MDL number MFCD00191145
  • Cas Number : 70897-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-119 �C
Synonym Z-L-Ser-NH2
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-serine-amide/5755?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-serine-amide/5755?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart