CH6371380670 Chem Impex International, Inc.

Z-3,5-diiodo-L-tyrosine ethyl ester

  • MDL number MFCD00191084
  • Cas Number : 102202-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-3,5-diiodo-L-Tyr-OEt
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-3-5-diiodo-l-tyrosine-ethyl-ester/5757?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-3-5-diiodo-l-tyrosine-ethyl-ester/5757?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart