CH6371380671 Chem Impex International, Inc.

Z-O-benzyl-L-tyrosine N-hydroxysuccinimide ester

  • MDL number MFCD00158646
  • Cas Number : 52773-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-144
Synonym Z-L-Tyr(Bzl)-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form Slightly cream colored powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-benzyl-l-tyrosine-i-n-i-hydroxysuccinimide-ester/5758?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-benzyl-l-tyrosine-i-n-i-hydroxysuccinimide-ester/5758?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart