CH6371380674 Chem Impex International, Inc.

Z-L-valine amide

  • MDL number MFCD00191170
  • Cas Number : 13139-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-212
Synonym Z-L-Val-NH2
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-valine-amide/5762?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-valine-amide/5762?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart