CH6371380675 Chem Impex International, Inc.

Z-Val-Ser-OH

  • MDL number MFCD00136594
  • Cas Number : 72635-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 175
Synonym Z-L-valyl-L-serine
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-val-ser-oh/5763?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-val-ser-oh/5763?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart