CH6371380677 Chem Impex International, Inc.

L-Homocystine bis-methyl ester dihydrochloride

  • MDL number MFCD06795760
  • Cas Number : 147857-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (H-L-HomoCys-OMe)2 2HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-homocystine-bis-methyl-ester-dihydrochloride/5765?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-homocystine-bis-methyl-ester-dihydrochloride/5765?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart