CH6371380684 Chem Impex International, Inc.

D-Leucine ethyl ester hydrochloride

  • MDL number MFCD00190936
  • Cas Number : 73913-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 127
Synonym D-Leu-OEt HCl
Purity 0.99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-leucine-ethyl-ester-hydrochloride/5773?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-leucine-ethyl-ester-hydrochloride/5773?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart