CH6371380690 Chem Impex International, Inc.

Fmoc-D-aspartic acid b-allyl ester

  • MDL number MFCD01074688
  • Cas Number : 177609-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Asp(OAll)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-aspartic-acid-beta-allyl-ester/5780?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-aspartic-acid-beta-allyl-ester/5780?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart