CH6371380691 Chem Impex International, Inc.

Fmoc-D-glutamic acid gamma-allyl ester

  • MDL number MFCD01074690
  • Cas Number : 204251-33-2
  • Molecular Weight : 409.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-131 °C
Purity 99.7
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-glutamic-acid-gamma-allyl-ester/5781?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-glutamic-acid-gamma-allyl-ester/5781?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart