CH6371380693 Chem Impex International, Inc.

Fmoc-O-allyl-D-tyrosine

  • MDL number MFCD01074692
  • Cas Number : 204269-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-151
Synonym Fmoc-D-Tyr(All)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-allyl-d-tyrosine/5783?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-allyl-d-tyrosine/5783?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart