CH6371380694 Chem Impex International, Inc.

Na-Fmoc-D-tryptophan N-hydroxysuccinimide ester

  • MDL number MFCD01076144
  • Cas Number : 174080-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-107 �dec.)
Synonym Fmoc-D-Trp-OSu
Purity 97
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-d-tryptophan-i-n-i-hydroxysuccinimide-ester/5784?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-d-tryptophan-i-n-i-hydroxysuccinimide-ester/5784?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart