CH6371380695 Chem Impex International, Inc.

Fmoc-3,5-dibromo-D-tyrosine

  • MDL number MFCD01076139
  • Cas Number : 204693-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-3,5-dibromo-D-tyr-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-5-dibromo-d-tyrosine/5785?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-5-dibromo-d-tyrosine/5785?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart