CH6371380697 Chem Impex International, Inc.

4-Cyano-DL-phenylalanine

  • MDL number MFCD01862353
  • Cas Number : 22888-47-7
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231-237 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-cyano-dl-phenylalanine/5787?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-cyano-dl-phenylalanine/5787?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart