CH6371380701 Chem Impex International, Inc.

Na-Fmoc-Ng-xanthyl-L-asparagine

  • MDL number MFCD07366857
  • Cas Number : 185031-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-220
Synonym Fmoc-L-Asn(Xan)-OH
Purity 96
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-xanthyl-l-asparagine/5791?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-xanthyl-l-asparagine/5791?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart