CH6371380702 Chem Impex International, Inc.

Nalpha-Boc-L-lysine methyl ester hydrochloride

  • MDL number MFCD00076957
  • Molecular Weight : 296.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at = -15 �C
Physical Form White to off-white solid (hygroscopic)
Shipping Temperature Ships with blue ice
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-l-lysine-methyl-ester-hydrochloride/5792?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-l-lysine-methyl-ester-hydrochloride/5792?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart