CH6371380703 Chem Impex International, Inc.

Benzyl-O-benzyl-L-serine methyl ester hydrochloride

  • MDL number MFCD02684385
  • Cas Number : 746598-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Light yellow oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-i-o-i-benzyl-l-serine-methyl-ester-hydrochloride/5793?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-i-o-i-benzyl-l-serine-methyl-ester-hydrochloride/5793?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart