CH6371380707 Chem Impex International, Inc.

N-Methyl-DL-aspartic acid hydrate

  • MDL number MFCD00150974
  • Cas Number : 303750-06-3
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-dl-aspartic-acid-hydrate/5797?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-dl-aspartic-acid-hydrate/5797?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart