CH6371380708 Chem Impex International, Inc.

N-Methyl-DL-tyrosine hydrochloride

  • MDL number MFCD06795796
  • Cas Number : 19897-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-180
Synonym N-Me-DL-Tyr-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-dl-tyrosine-hydrochloride/5798?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-dl-tyrosine-hydrochloride/5798?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart