CH6371380712 Chem Impex International, Inc.

Na-Fmoc-Ng-Z-D-2,4-diaminobutyric acid

  • MDL number MFCD02684372
  • Cas Number : 387824-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-133
Synonym Fmoc-D-Dab(Z)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-z-d-2-4-diaminobutyric-acid/5802?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-z-d-2-4-diaminobutyric-acid/5802?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart