CH6371380714 Chem Impex International, Inc.

Fmoc-D-glutamic acid a-allyl ester

  • MDL number MFCD01074691
  • Cas Number : 204251-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-128
Synonym Fmoc-D-Glu-OAll
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-glutamic-acid-alpha-allyl-ester/5804?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-glutamic-acid-alpha-allyl-ester/5804?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart