CH6371380719 Chem Impex International, Inc.

Fmoc-L-cysteine amide

  • MDL number MFCD03788160
  • Cas Number : 623177-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-cysteine-amide/5829?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-cysteine-amide/5829?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart