CH6371380720 Chem Impex International, Inc.

Nalpha-Fmoc-Nbeta-(N-Boc-amino-oxyacetyl)-L-2,3-diaminopropionic acid

  • MDL number MFCD02682868
  • Cas Number : 600153-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-136 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-beta-sup-i-n-i-boc-amino-oxyacetyl-l-2-3-diaminopropionic-acid/5830?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-beta-sup-i-n-i-boc-amino-oxyacetyl-l-2-3-diaminopropionic-acid/5830?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart