CH6371380726 Chem Impex International, Inc.

Fmoc-(1-Boc-piperidin-3-yl)-DL-glycine

  • MDL number MFCD02683126
  • Cas Number : 494210-72-9
  • Molecular Weight : 480.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-1-boc-piperidin-3-yl-dl-glycine/5840?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-1-boc-piperidin-3-yl-dl-glycine/5840?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart