CH6371380729 Chem Impex International, Inc.

Fmoc-N-phenylmethyl-hydroxylamine

  • MDL number MFCD02682860
  • Cas Number : 198411-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-n-phenylmethyl-hydroxylamine/5844?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-n-phenylmethyl-hydroxylamine/5844?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart