CH6371380735 Chem Impex International, Inc.

O-(N-Boc-2-aminoethyl)-O'-(N-diglycolyl-2-aminoethyl)hexaethyleneglycol

  • MDL number MFCD09859788
  • Cas Number : 600141-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-NH-(PEG)6-COOH (30 atoms)
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white to yellow oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/o-n-boc-2-aminoethyl-o-n-diglycolyl-2-aminoethyl-hexaethyleneglycol/5850?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/o-n-boc-2-aminoethyl-o-n-diglycolyl-2-aminoethyl-hexaethyleneglycol/5850?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart