CH6371380739 Chem Impex International, Inc.

Weinreb linker

  • MDL number MFCD01861163
  • Cas Number : 247021-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-N-methoxy-3-aminopropionic acid
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/weinreb-linker/5855?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/weinreb-linker/5855?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart