CH6371380763 Chem Impex International, Inc.

L-b-Homoglutamine hydrochloride

  • MDL number MFCD01862846
  • Cas Number : 336182-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-beta-HomoGln-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-beta-homoglutamine-hydrochloride/5884?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-beta-homoglutamine-hydrochloride/5884?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart