CH6371380764 Chem Impex International, Inc.

(S)-3-Aminoadipic acid hydrochloride

  • MDL number MFCD01862872
  • Cas Number : 61884-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym beta-HomoGlu-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-aminoadipic-acid-hydrochloride/5885?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-aminoadipic-acid-hydrochloride/5885?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart